missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) Sulfate, Technical, Fisher Chemical™

Catalog Number p-7073101
Click to view available options
Quantity:
500 g
1 kg
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
15244-10-7
Fe2H2O13S3
417.87
149714
VXWSFRMTBJZULV-UHFFFAOYSA-H
167265
diiron(3+) hydrate trisulfate
O.[Fe+3].[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
15244-10-7
Fe2H2O13S3
417.87
149714
VXWSFRMTBJZULV-UHFFFAOYSA-H
167265
diiron(3+) hydrate trisulfate
O.[Fe+3].[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Physical Form Solid
Quantity 500 g
Formula Weight 399.88g/mol
Packaging Plastic powder jar
compliance-icons compliance-icons
Product Identifier
  • Iron(III)sulfate hydrate
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contains : Sulfuric acid, iron(3+) salt (3:2), hydrate, Sulfuric acid,Ferric sulfate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'