missing translation for 'onlineSavingsMsg'
Learn More

Iso-Butyl Alcohol, Extra Pure, SLR, Fisher Chemical™

Catalog Number p-7072881
Click to view available options
Quantity:
2.5 L
25 L
Packaging:
Metal can
Plastic Coated Glass bottle
This item is not returnable. View return policy
78-83-1
C4H10O
4740
This item is not returnable. View return policy
78-83-1
C4H10O
4740
Melting Point -108°C
Color Colorless
Boiling Point 108°C
Packaging Plastic Coated Glass bottle
Quantity 2.5 L
Vapor Density 2.6
Formula Weight 74.12g/mol
Vapor Pressure 11.7mbar at 20°C
Physical Form Liquid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'