missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Isonipecotic Acid, 99%

Catalog Number p-3737953
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
498-94-2
C6H11NO2
129.16
MFCD00006004
SRJOCJYGOFTFLH-UHFFFAOYSA-N
isonipecotic acid, 4-piperidinecarboxylic acid, 4-carboxypiperidine, hexahydroisonicotinic acid, dl-isopipecolinic acid, isonicotinic acid, hexahydro, h-inp-oh, acide isonipecotique, 4-hexahydroisonicotinic acid, unii-m5tzp1rwie
3773
piperidine-4-carboxylic acid
OC(=O)C1CCNCC1
This item is not returnable. View return policy
498-94-2
C6H11NO2
129.16
MFCD00006004
SRJOCJYGOFTFLH-UHFFFAOYSA-N
isonipecotic acid, 4-piperidinecarboxylic acid, 4-carboxypiperidine, hexahydroisonicotinic acid, dl-isopipecolinic acid, isonicotinic acid, hexahydro, h-inp-oh, acide isonipecotique, 4-hexahydroisonicotinic acid, unii-m5tzp1rwie
3773
piperidine-4-carboxylic acid
OC(=O)C1CCNCC1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point >300.0°C
Color Beige-Pink to White
Loss on Drying 1% max. (120°C, 4 hrs)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 25 g
Beilstein 22, V,1, 244
Merck Index 15, 5235
Solubility Information (10% in water) Clear to slightly hazy
Formula Weight 129.16
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Isonipecotic acid
Show More Show Less
compliance-icons
Product Identifier
  • Isonipecotic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'