missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl-β-D-thiogalactopyranoside (IPTG) (Wht. Powd., Dioxane Crystalline), Fisher BioReagents™

Catalog Number p-26627
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
367-93-1
C9H18O5S
238.30
MFCD00063273
BPHPUYQFMNQIOC-NXRLNHOXSA-N
dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin
31275
CHEBI:47032
1,4-dioxane
CC(C)S[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O
This item is not returnable. View return policy
367-93-1
C9H18O5S
238.30
MFCD00063273
BPHPUYQFMNQIOC-NXRLNHOXSA-N
dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin
31275
CHEBI:47032
1,4-dioxane
CC(C)S[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O
Melting Point 122°C
Color White
Quantity 1 g
Packaging Glass Bottle
Physical Form Solid
Merck Index 15, 5126
Formula Weight 238.3
Chemical Name or Material Isopropyl-β-D-thiogalactopyranoside
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'