Learn More
Justrite™ Drum Faucets, Extender, and Valve
Self-closing safety drum faucets
Brand:  Justrite™ 08910
Code : 03
Additional Details : Weight : 0.52000kg
 View more versions of this product
Product Code. 11963549
  • 81.05 € / Each
Estimated Shipment: 04-06-2024
to see stock.
Add to basket
Description
Description
  • Self-closing brass safety faucets feature a PTFE fluorocarbon resin seal, flame arrester and standard 0.75 in. NPT thread
  • Brass faucets screw into drum; adjustable model adjusts to spout-down position
  • Flexible faucet extensions provide accuracy, prevent spills; available in 8 in. long (20.3cm) brass and 6 in. long (15.2cm) stainless steel
  • Brass drum gate valve fits 2 in. (5.1cm) NPT openings and has wide mouth for easy dispensing of viscous liquids from drums in horizontal position
Certifications
Faucets and valve are FM approved.
Specifications
Specifications
Adjustable Drum Faucet
FM
Brass
PTFE Fluorocarbon Resin seal, internal flame arrester and