missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ Flammables Single-Door Benchtop Cabinet

Allows every lab to have a small flammables storage cabinet

Brand:  Justrite™ 8904001

Code : 03

Additional Details : Weight : 28.12272kg

Product Code. 12351683

  • 852.00 € / Each
Estimated Shipment: 20-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Includes: One adjustable shelf

Description

Description

Ideal for localized storage of small quantities of flammable laboratory chemicals.

Features of Justrite Flammables Cabinets

  • Double-wall construction provides 1.5 in. of insulating air space to help prevent the spread of flames
  • High-visibility Haz-Alert™ reflective warning labels offer added safety during power outages or fires
  • Dual vents with flame arresters at top and opposite bottom allow hazardous vapors to be piped away
  • Easy-closing, self-latching doors facilitate fire protection of the cabinet
  • Self latch engages critical three-point latching system for protection under fire conditions
  • SpillSlope™ safety shelves direct incidental spills to the back and bottom of leakproof sump
  • Exterior side panel has built-in grounding connector for easy grounding
  • Finished inside and out with durable lead-free powder paint
  • 1.5 in. (3.8cm) air space between 18G steel walls
  • 2 in. (5cm) vent openings
  • With manual closing door

Order Info

Capacity: 4 gal. (15L). Dimensions: 17L × 17W × 22 in.H (43 × 43 × 56cm). Weight: 62 lb. (28kg).

Compliance

Meet OSHA and NFPA requirements

Services

10-Year

Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers