missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ Biohazard Waste Can

The Justrite Biohazard Waste Can is a sturdy, steel can used to store contaminated laundry as well as other regulated waste. Can features a leakproof construction with foot-operated, self-closing lid.

Brand:  Justrite™ 05910

Code : NEW

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11934977

  • 158.00 € / Each
Estimated Shipment: 31-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Safely segregate biohazard waste to ensure proper disposal of blood- or pathogen-contaminated laundry items and pathological and microbiological wastes.

  • Features leakproof, sturdy steel construction with foot-operated, self-closing lid
  • Eliminates any reason to reach into the waste area or touch the can
  • Holds biohazard bag (sold separately) securely around circumference of can top
  • Large biohazard label clearly identifies the contents and alerts workers

Not to be used for disposal of sharp items.

Specifications

Specifications

Biohazard Waste Container
FM, TUV, OSHA
12 in.
White
Foot-operated cover
16 in.
Metal
30.2 cm
6 gal.
40.64 cm
Safe, temporary collection of biological waste
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers