missing translation for 'onlineSavingsMsg'
Learn More

K-252a, 98%, , Thermo Scientific Chemicals

Catalog Number p-152579
Click to view available options
Quantity:
1 mg
100 μg
This item is not returnable. View return policy
97161-97-2
C27H21N3O5
467.5
MFCD00161522
KOZFSFOOLUUIGY-UHFFFAOYSA-N
methyl 6-hydroxy-5-methyl-13-oxo-5,6,7,8,14,15-hexahydro-13h-5,8-epoxy-4b,8a,14-triazadibenzo b,h cycloocta 1,2,3,4-jkl cyclopenta e-as-indacene-6-carboxylate
3813
CC12C(CC(O1)N3C4=CC=CC=C4C5=C6C(=C7C8=CC=CC=C8N2C7=C53)CNC6=O)(C(=O)OC)O
This item is not returnable. View return policy
97161-97-2
C27H21N3O5
467.5
MFCD00161522
KOZFSFOOLUUIGY-UHFFFAOYSA-N
methyl 6-hydroxy-5-methyl-13-oxo-5,6,7,8,14,15-hexahydro-13h-5,8-epoxy-4b,8a,14-triazadibenzo b,h cycloocta 1,2,3,4-jkl cyclopenta e-as-indacene-6-carboxylate
3813
CC12C(CC(O1)N3C4=CC=CC=C4C5=C6C(=C7C8=CC=CC=C8N2C7=C53)CNC6=O)(C(=O)OC)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Assay Percent Range 97.5% min. (HPLC)
Quantity 100 μg
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in dichloromethane,dmso
Formula Weight 467.5
Percent Purity 98%
Chemical Name or Material K-252a

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'