missing translation for 'onlineSavingsMsg'
Learn More

Kir6.1 (KCNJ8) Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA548354

Additional Details : Weight : 0.01000kg

Product Code. 15622655

  • 473.00 € / 400µL
Estimated Shipment: 18-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Predicted to react with mouse and rat based on sequence homology.

Kir6.1 (KCNJ8) Polyclonal antibody specifically detects Kir6.1 (KCNJ8) in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.
Specifications

Specifications

Kir6.1 (KCNJ8)
Polyclonal
PBS with 0.09% sodium azide
Q15842
KCNJ8
KLH conjugated synthetic peptide between 4-33 amino acids from the N-terminal region of human KCNJ8.
400 μL
Primary
Human
Antibody
IgG
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
KCNJ8
AI448900; ATP-sensitive inward rectifier potassium channel 8; ATP-sensitive inward rectifier potassium channel Kir6.1; gnite; Inward rectifier K(+) channel Kir6.1; inward rectifier potassium channel 6.1; inward rectifier potassium channel Kir6.1; inwardly rectifier K(+) channel Kir6.1; Inwardly rectifying potassium channel gene subfamily J-8 (ATP sensitive); Inwardly rectifying potassium channel gene, subfamily J-8 (ATP sensitive); inwardly rectifying potassium channel Kir6.1; ion channel; Kcnj8; KCNJ8 protein; Kir6.1; Kir6.1 like; potassium channel, inwardly rectifying subfamily J member 8; potassium channel, inwardly rectifying subfamily J, member 8; potassium inwardly rectifying channel subfamily J member 8; potassium inwardly-rectifying channel J8; potassium inwardly-rectifying channel, subfamily J, member 8; potassium voltage-gated channel subfamily J member 8; si:dkey-183c2.2; slmbr; sltr; UKATP1; uKATP-1; zkir6.1
Rabbit
Antigen affinity chromatography, Protein A
RUO
3764
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers