missing translation for 'onlineSavingsMsg'
Learn More

KD Scientific Stainless-Steel Syringes

Durable, electron-beam construction withstands high force, high pressure applications

Brand:  KD Scientific 780805

Code : A0

Additional Details : Weight : 0.30000kg

 View more versions of this product

Product Code. 11945596

  • 1435.00 € / Each
Estimated Shipment: 08-08-2024
to see stock.
Add to basket

Description

Description

Eliminates hazards of glass syringe breakage.

  • Compatible with most syringe pumps
  • Fully autoclavable for reuse
  • Adaptable to Luer Lock or Swagelok™ fittings
  • Resistant to most chemicals

Specifications

Specifications

100 mL
1-1/5 in.
34.7 mm
Stainless Steel (316)
1500 psi
1-37/100 in.
38.1 mm
Syringe Pump
Swagelok 1/16 in. Syringe
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers