Learn More
Lenz Laborglasintrumente™ Keck™ Conical Joint Clips
POM clip for conical ground joints
Brand: Lenz Laborglasintrumente™ 01103734
Additional Details : Weight : 0.60000kg
Description
- Outstanding chemical resistance
- Operating temperature up to approximately 150 °C
Specifications
10 | |
For ground joint NS 34/35 | |
Orange |