missing translation for 'onlineSavingsMsg'
Learn More

Ketoconazole, 98%, Thermo Scientific™

Potent inhibitor of cytochrome P450c17

165.00 € - 566.00 €


Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15415619
SDS View Documents
Thermo Scientific Alfa Aesar
J63367.06
165.00 €
5g
View alternative products
Save Up to 
»
In Stock
Add to basket
 
 Restricted Item
15425619
SDS View Documents
Thermo Scientific Alfa Aesar
J63367.14
25g
566.00 €
25g
View alternative products
Save Up to 
»
Estimated Shipment: 22-09-2026
Add to basket
 
 Restricted Item
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Ketoconazole is used to treat candidiasis, chronic mucocutaneous candidiasis, oral thrush, candiduria, blastomycosis, coccidioidomycosis, histoplasmosis, chromomycosis, and paracoccidioidomycosis. Ketoconazole is an antifungal agent.

Solubility
Soluble in DMSO, ethanol, chloroform, water, and methanol.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Videos
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.