Learn More
KIAA0895L Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA564255
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: EDVDHLRPHG VLDNTRVPHF MQDLARYRQQ LEHIMATNRL DEAELGRLLP D Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 88%.
KIAA0895L Polyclonal antibody specifically detects KIAA0895L in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
KIAA0895L | |
Polyclonal | |
Unconjugated | |
KIAA0895L | |
4931428F04Rik; AI426165; hypothetical protein LOC688736; KIAA0895 like; Kiaa0895l; KIAA0895-like; KIAA0895-like ortholog; RIKEN cDNA 4931428F04 gene; uncharacterized protein KIAA0895-like; uncharacterized protein KIAA0895-like homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
653319 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q68EN5 | |
KIAA0895L | |
Recombinant protein corresponding to Human KIAA0895L | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only