missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kimberly-Clark Professional™ KleenGuard™ G80 Nitrile Chemical Resistant Gloves
Green, hand-specific gloves provide maximum protection against chemical and mechanical hazards. G80 Chemical Resistant Latex-Free Gloves include improved, more consistent cotton flock lining for added comfort.
50.60 € - 216.00 €
Specifications
Certifications/Compliance | FDA, ASTM D3767, ASTM D412, ASTM D5151, ASTM D6319 |
---|---|
Chlorinated | No |
Color | Green |
Latex Free | Yes |
Material | Nitrile |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
11730654
|
Kimberly-Clark Professional™
94445 |
Small |
50.60 €
12 pairs |
In Stock
Log in to see stock. |
|||||
11740654
|
Kimberly-Clark Professional™
94446 |
Medium |
50.60 €
12 pairs |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
11750654
|
Kimberly-Clark Professional™
94447 |
Large |
50.60 €
12 pairs |
Estimated Shipment: 23-05-2024
Log in to see stock. |
|||||
11770654
|
Kimberly-Clark Professional™
94448 |
X-Large |
50.60 €
12 pairs |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||||
11780654
|
Kimberly-Clark Professional™
94449 |
2X-Large |
50.60 €
12 pairs |
Estimated Shipment: 20-08-2024
Log in to see stock. |
|||||
12691486
|
Kimberly-Clark Professional™
94446 |
Medium |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15326758
|
Kimberly-Clark Professional™
94445 |
Small |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12601496
|
Kimberly-Clark Professional™
94447 |
Large |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15386758
|
Kimberly-Clark Professional™
94447 |
Large |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15376758
|
Kimberly-Clark Professional™
94446 |
Medium |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12611496
|
Kimberly-Clark Professional™
94448 |
X-Large |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15306768
|
Kimberly-Clark Professional™
94448 |
X-Large |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15316768
|
Kimberly-Clark Professional™
94449 |
2X-Large |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12621496
|
Kimberly-Clark Professional™
94449 |
2X-Large |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12681486
|
Kimberly-Clark Professional™
94445 |
Small |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Hand specific latex-free gloves protect hands against chemical or mechanical hazards
- Diamond embossed fingertips offer enhanced wet and dry grip
- Flock-lined for easy donning and extra comfort in extended use
- Improved AQL of 0.65 versus 1.5, resulting in fewer pinholes
- Modified Nitrile formulation for more durability and enhanced chemical resistance.
- Now available in 5 bulk-packed poly bags versus individually wrapped pairs
- Ideal for: providing PPE category 3 protection, level 4 abrasion resistance and durability in manufacturing, transport, construction, agriculture cleaning and janitorial work, when handling chemicals, oils, grease, acids, caustics or solvents
Specifications
FDA, ASTM D3767, ASTM D412, ASTM D5151, ASTM D6319 | |
Green | |
Nitrile | |
0.38 mm | |
No | |
JKLNOP | |
Acids, bases, Solvents and Caustics | |
Cotton Flock | |
KleenGuard™ G80 | |
33 cm |
No | |
Yes | |
15 mil | |
EN 388:4101X | |
Type A | |
Bacteria, Fungi only | |
Rough | |
Lined | |
13 in. |