Learn More
DWK Life Sciences Kimble™ AmberScrew Thread Vials with Attached White Rubber Lined Closures
Attached closed top black phenolic closures
POA
Specifications
Material (Liner) | Rubber |
---|---|
Color | Amber |
Material | Borosilicate Glass |
Thread Style | Screw |
Closure Material | Phenolic |
Description
Attached closed top black phenolic closures
- Manufactured from amber 51 expansion borosilicate glass
- Closures with white rubber liners
- Vials packaged in corrugated trays with partitions
Specifications
Rubber | |
Borosilicate Glass | |
Phenolic |
Amber | |
Screw | |
USP Type I |