Learn More
DWK Life Sciences Kimble™ Clear Screw Thread Sample Vials without Closures
Packaged in corrugated trays with partitions
POA
Specifications
Color | Clear |
---|---|
Material | Borosilicate Glass |
Thread Style | Screw |
Certifications/Compliance | ASTM Type I, Class A |
Description
- Manufactured from clear 33 expansion borosilicate glass
Specifications
Clear | |
Borosilicate Glass | |
Screw | |
ASTM Type I, Class A |