Learn More
DWK Life Sciences Kimble™ KIMAX™ Colorware™ Flasks
Colorful enamel graduations enhance readability and indicate approximate volumes
POA
Specifications
Closure Material | Rubber |
---|---|
Material | Borosilicate Glass |
Writing Surface | Matte Finish Marking |
Mouth Style | Narrow |
Description
Colorful enamel graduations enhance readability and indicate approximate volumes
- Narrow mouth
- Durable, matte-finish marking spot provides good writing surface
- Without stoppers
Compliance
Designed from ASTM™ E 1404, Type I, Class 1 requirements
Specifications
Rubber | |
Borosilicate Glass | |
Matte Finish Marking | |
Narrow |