Learn More
DWK Life Sciences Kimble™ KIMAX™ Flasks with Quick-Release Hose Connector
Allow safe attachment of 0.25 in. I.D. (6mm) tubing
POA
Specifications
Material Borosilicate Glass
View More Specs
Description
Description
Allow safe attachment of 0.25 in. I.D. (6mm) tubing
  • Tubing remains permanently attached to hose barb and opposite end attaches to flask tubulation with positive threaded seal
  • Proper positioning of connector provides greater flask stability as tubing angled downward has less tendency to tilt flask
  • Graduated with approximate volume scale in durable white ceramic enamel
  • Tubing and stoppers sold separately
Specifications
Specifications
Borosilicate Glass