missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kimble Chase TLC tank cylindrical with polyethylene lid glass for 1 inch x 3 inch plates Kimble
Brand: Kimble Chase 4161701118
TLC tank cylindrical with polyethylene lid glass for 1 inch x 3 inch plates Kimble
Code : 16
Additional Details : Weight : 0.84600kg