missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Kontes™ Micro-Vial Graduated Screw Thread Vials

With solid-top closure and PTFE-faced white rubber liner

Brand:  DWK Life Sciences 60702-1

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10010802

  • 142.00 € / Pack of 12


Estimated Shipment: 22-09-2021
Add to basket

Description

Description

  • Accuform™
  • KG-33 borosilicate glass
  • Extra thick clear glass walls

Specifications

Specifications

Clear
Borosilicate Glass
Yes
PTFE/Rubber
13-425
49 mm
14 mm
1 mL
1.92 in.
Tapered
Screw Cap (Open-Top)
12 Case
Videos
SDS
Documents
Certificates