missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Kontes™ Packing Reservoir for CHROMAFLEX™ Columns

CHROMAFLEX™ packing reservoirs are ideal for packing gel slurries into columns

Brand:  DWK Life Sciences 420837-0040

Code : NEW

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 15320168

  • 1420.15 € / Each


Estimated Shipment
06-11-2020
Add to basket

Description

Description

  • Fits Standard Column
  • Each packing reservoir is designed for easy mounting on the CHROMAFLEX™ columns
  • The capacities are matched to take the amount of gel slurry required to pack the longest column
  • The packing reservoirs may also be used as buffer reservoirs for gravity flow chromatography
  • 1.0cmID column has nominal volume of 150mL
  • 2.5cmID column has nominal volume of 500mL
  • Temperature Range 0 to 50°C
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications

Specifications

4.8 cm
End fitting has 1/4-28 threads for easy connection to pumps
2000 mL
Videos
Documents