missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ Kontes™ Pestle For All-Glass DUALL™ Tissue Grinders
Grinding efficiency is greatly improved when this tube is used. Available in sizes 20 - 23.
139.00 €
Specifications
Product Type | Tissue grinder pestle |
---|---|
For Use With (Equipment) | Duall™ all Glass tissue grinders |
Description
Grinding efficiency is greatly improved when this tube is used. Available in sizes 20 - 23.
- Combines both conical and cylindrical surfaces to effectively reduce tissue and produce a uniform homogenate.
- This ground glass model is ideal for connective tissue such as muscle, heart and lung
- Construction is strong enough to allow the pestle to be motor-driven
- Clearance between pestles and tubes is 0.004 in. to 0.006 in.
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Tissue grinder pestle | |
Duall™ all Glass tissue grinders |