Learn More
DWK Life Sciences Kimble™ Kontes™ Chromatography Column Accessories
For chromatography columns
Brand: DWK Life Sciences 747300-0050
Additional Details : Weight : 0.09000kg
Description
- 747000 Series Microflex Kits
- Features versatile threaded chaste connections
- Can be assembled rightside up or upside down, providing great versatility
- GPI thread size: 13 to 425
Specifications
Drying Tube/Chromatography Column | |
For chromatography columns | |
13-425 |