missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ Linear HDPE Stoppers
Closed bottom end fits the standard taper neck
40.90 €
Specifications
Color | Yellow, Blue |
---|---|
Material | HDPE |
Product Type | Stopper |
For Use With (Application) | Used as replacement parts for items that are ground to standard taper stopper dimensions |
Description
Closed bottom end fits the standard taper neck
- Heavy-duty Polyethylene stopper
- Flange on stopper protects glass
Specifications
Yellow, Blue | |
HDPE | |
Stopper | |
Used as replacement parts for items that are ground to standard taper stopper dimensions |