missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Kinase buffer II (5X)
For cellular and molecular biology applications
44.80 € - 65.60 €
Specifications
Components | 100mM MOPS, 50mM magnesium acetate, 5mM DTT 5, 5mM EDTA, 25mM beta glycerophosphate, and 5mM sodium orthovanadate, pH 7.4 |
---|---|
Content And Storage | Ambient temperatures |
Product Type | Kinase buffer II (5X) |
Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15463059
|
Thermo Scientific™
J60269.AE |
100 mL |
44.80 €
100mL |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15473059
|
Thermo Scientific™
J60269.AK |
250 mL |
65.60 €
250mL |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Formulation: 100mM MOPS, 50mM magnesium acetate, 5mM DTT 5, 5mM EDTA, 25mM beta glycerophosphate, and 5mM sodium orthovanadate, pH 7.4
Specifications
100mM MOPS, 50mM magnesium acetate, 5mM DTT 5, 5mM EDTA, 25mM beta glycerophosphate, and 5mM sodium orthovanadate, pH 7.4 | |
Ambient temperatures | |
Kinase buffer II (5X) | |
Liquid |
For Research Use Only