missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ KingFisher Apex Purification Systems

The KingFisher Apex Purification System is a versatile and user-friendly instrument that uses magnetic particle processing to automate the purification or isolation of DNA, RNA, proteins, and cells from virtually any source.

Brand:  Thermo Scientific™ 5400930


Additional Details : Weight : 56.00000kg

 View more versions of this product

Product Code. 16512452

  • POA / Each
Estimated Shipment: 13-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.


Description

Description

The KingFisher Apex Purification System is a versatile and user-friendly instrument that uses magnetic particle processing to automate the purification or isolation of DNA, RNA, proteins, and cells from virtually any source. It moves magnetic beads, not liquids, through the main steps of sample preparation: bind, wash, and elute. This process provides excellent quality, reproducible results, and reduces hands-on time for sample processing.

  • User-friendly high throughput sample purification system for automating DNA/RNA, protein and cell purification
  • Provides versatile and high-speed purification with reproducible results
  • Easy to use interface with touch screen empowering the user to control all details of the application and purification process
  • BindIx PC Software for added convenience
  • Extensive selection of validated protocols for different type of applications
  • Cloud library for quick protocol retrieval
Specifications

Specifications

Plasma, Blood, Urine, Saliva, Cell Homogenate, Tissue Homogenate
96 KF plate 50 to 150 μL, 96 DW plate 50 to 1000 μL
96-well
KingFisher Apex with 96 DW head
Down to +4°C
Genomics and proteomics, target identification, veterinary assays, biomarker discovery, quality control, forensics
96 in one frame, interchangeable
96
Max 23 hr. 59 min.
96 KF, 96 DW
56 cm
46 cm
56 kg
79 cm
PCR/qPCR, Sequencing, Mass Spec
8
96 Samples
>98% with 2.8 μm particle size
Touch screen (10 in.)
+4°C above ambient temperature up to +100°C
ca. >1 μm
96 in one frame, interchangeable
2 pcs
22 in.
18 in.
123 lb.
31 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Services