Learn More
Thermo Scientific™ KingFisher™ Pure DNA Blood Kit
Achieve rapid, reproducible purification of high-quality DNA from fresh or frozen whole blood treated with EDTA or citrate with this unique nucleic acid purification workflow.
POA
Specifications
Sample Type | Fresh or frozen whole blood treated with EDTA or citrate |
---|---|
Suitable Downstream Analysis | PCR, Real-time PCR, Enzymatic Reactions |
Elution Volume (Metric) | 200µL sample: 100µL (Duo) or 150µL (Flex). 1mL sample: 500µL. Recommended but adjustable |
Description
Achieve rapid, reproducible purification of high-quality DNA. The Thermo Scientific™ KingFisher™ Pure DNA Blood Kit, together with a KingFisher magnetic particle processor, allows rapid and reproducible purification of high-quality DNA from fresh or frozen whole blood treated with EDTA or citrate. Thermo Scientific KingFisher Kits complete the unique nucleic acid purification workflow providing an optimized high throughput method for extreme flexibility.
Features- Produces isolated DNA that is free of inhibitors and ready for use in many downstream applications
- Easy-to-use protocols for safe sample-handling, as well as minimal user input, maximizing walkaway time.
- Excellent performance and consistency of results optimized to produce high yields of quality DNA
- Large volume sample input possible up to 3mL of blood per sample
- Flexible throughput from one sample to up to 96 samples per run
- Minimized hands-on time increases efficiency
- Compatible with most common downstream analysis and applications
Recommended for:
Genomics, target identification, veterinary assays, biomarker discovery, quality control
Specifications
Fresh or frozen whole blood treated with EDTA or citrate | |
PCR, Real-time PCR, Enzymatic Reactions | |
200µL sample: 100µL (Duo) or 150µL (Flex). 1mL sample: 500µL. Recommended but adjustable |