Learn More
Kjeldahl Copper Catalyst Tablets, Technical, Contains 1g Na2SO4 And Equivalent Of 0.1g CuSO4, Fisher Chemical™
Kjeldahl Copper Catalyst Tablets, Technical, Contains 1g Na2SO4 And Equivalent Of 0.1g CuSO4, Fisher Chemical, Quantity: 1000 tablets, Blue-White, MDL Number: 147336, Packaging: Plastic powder jar, Physical Form: Solid
Brand: Fisher Chemical K/0120/81
Code : 21
Additional Details : CAS Number : 7757-82-6 Weight : 1.23000kg Transport : UN number : 3077 Chem class : 9 Pack group : III
Description
Kjeldahl Copper Catalyst Tablets, Technical, Contains 1g Na2SO4 And Equivalent Of 0.1g CuSO4, Fisher Chemical, Quantity: 1000 tablets, Blue-White, MDL Number: 147336, Packaging: Plastic powder jar, Physical Form: Solid
Specifications
Plastic powder jar | |
1000 tablets | |
Solid |
Blue-White | |
147336 |