missing translation for 'onlineSavingsMsg'
Learn More

Ecolab™ Klerwipe™ Sterile Biocide Impregnated Wipes

For use in rapid clean down of surfaces, equipment and production components. Ecolab™ Klerwipe™ Sterile Biocide Impregnated Wipes remain wet throughout use ensuring consistent application of alcohol to whole surface area.

Brand:  Ecolab™ 3079290

Code : NEW

Additional Details : Weight : 6.50000kg

 View more versions of this product

Product Code. 15326128

  • 324.00 € / Pack of 20


UK Stock 3

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Effective biocides including good sporicidal activity
  • Rapid and effective application
  • Polythene packaging features Klertear which makes it easy to open even when wearing gloves
  • Non-toxic and non-corrosive
  • Ideal for sterile environments
  • Either gamma irradiated, or aseptically processed
  • Each pouch is supplied double bagged
  • No wastage

Certifications

Manufactured by ISO 9001:2008, ISO 13485:2012 and European Medical Device Directive 93/42/EEC accredited company

Specifications

Specifications

Highly absorbent
Rapid clean down of surfaces, equipment and production components
Sterile impregnated wipes
200 x 200mm (7.8 x 7.8 in.)
Cellulose, Polyester
With Sporicidal Chlorine/Quat
Videos
Documents