missing translation for 'onlineSavingsMsg'
Learn More

BINDER™ KMF Alarm Output

Binder™ KMF Alarm Output is an accessory for use with a variety of Humidity Test Chambers.

Brand:  BINDER™ 8012-1676

Additional Details : Weight : 0.20000kg

Product Code. 15626527

  • 1690.00 € / Each
Estimated Shipment: 28-05-2024
to see stock.
Add to basket

Description

Description

  • Alarm output
  • Zero voltage
  • Humidity (±5% RH)
  • Accessible via 6-pin DIN socket
  • Features audible signal that can be switched off
Specifications

Specifications

Alarm output, zero-voltage - for temperature (±2°C) and humidity (±5 % RH), accessible via 6-pin DIN socket (max. 24 V - 2.5 A), with audible signal that can be switched off
BINDER Chambers KMF 115
2.5 A
2°C
Audible signal that can be switched off
24 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers