Learn More
DWK Life Sciences Kimble™ Kontes™ Brand Rotary Evaporator Glass Vial Connecting Adapters
Ideal for use with small samples
POA
Description
Ideal for use with small samples
Connect standard rotary evaporator traps to vials with variety of GPI thread sizes. Typical receivers include Accuform™ 1 to 10mL vials (06-100C, 06-100E, 06-100F, 06-100G). Any standard vial with GPI threads can be used.
Specifications
Glass |