Learn More
DWK Life Sciences Kimble™ Kontes™ Squibb Pear-Shaped Separatory Funnels with standard taper Stopper and Autoclavable PTFE Stockcock Plug
Supplied with a standard taper ground glass stopper and a totally autoclavable PTFE stopcock plug
POA
Specifications
Material | Borosilicate Glass |
---|---|
Certifications/Compliance | ASTM E1096, Type IV |
Length (Metric) Stem | 60mm |
Stopper Material | Glass |
Product Type | Squibb separatory funnel |
Description
Kimble Chase KimCote™ plastic-coated glassware provides an added measure of safety in the event of breakage
- Lower stems have inside diameter large enough so that a column of liquid will break with the stopcock closed, thereby giving a more complete separation and eliminating the necessity of emptying the funnel
- Designed from ASTM™ Specifications E1096, Type 4 requirements
Specifications
Borosilicate Glass | |
60mm | |
Squibb separatory funnel |
ASTM E1096, Type IV | |
Glass |