Learn More
L-3,4-Dihydroxyphenylalanine methyl ester hydrochloride, 95%, ACROS Organics™
Brand: Acros Organics 439490050
Code : 29
Additional Details : CAS Number : 1421-65-4 Weight : 0.05500kg
Quantity | 5g |
---|
Chemical Identifiers
1421-65-4 | |
247.68 | |
s-methyl 2-amino-3-3,4-dihydroxyphenyl propanoate hydrochloride, l-3,4-dihydroxyphenylalanine methyl ester hydrochloride, methyl l-dopa hydrochloride, melevodopa hydrochloride, unii-jfu1a8866v, l-dopa methyl ester hydrochloride, levodopa methyl ester hydrochloride, l-tyrosine, 3-hydroxy-, methyl ester, hydrochloride, dsstox_cid_25763, dsstox_rid_81105 | |
methyl (2S)-2-amino-3-(3,4-dihydroxyphenyl)propanoate;hydrochloride |
C10H13NO4·ClH | |
WFGNJLMSYIJWII-FJXQXJEOSA-N | |
10131132 | |
COC(=O)C(CC1=CC(=C(C=C1)O)O)N.Cl |
Specifications
L-3,4-Dihydroxyphenylalanine methyl ester hydrochloride | |
Authentic | |
+9.0° to +11.0° (20°C, 589nm) (C=2 in methanol) | |
White | |
5g | |
95% | |
s-methyl 2-amino-3-3,4-dihydroxyphenyl propanoate hydrochloride, l-3,4-dihydroxyphenylalanine methyl ester hydrochloride, methyl l-dopa hydrochloride, melevodopa hydrochloride, unii-jfu1a8866v, l-dopa methyl ester hydrochloride, levodopa methyl ester hydrochloride, l-tyrosine, 3-hydroxy-, methyl ester, hydrochloride, dsstox_cid_25763, dsstox_rid_81105 | |
COC(=O)C(CC1=CC(=C(C=C1)O)O)N.Cl | |
247.68 | |
247.68 | |
95% |
45.58 to 51.40 % (C) Dry basis 5.32 to 5.99 % (N) Dry basis | |
Glass Bottle | |
1% Max. | |
94.2°C to 94.3°C | |
1421-65-4 | |
C10H13NO4·ClH | |
WFGNJLMSYIJWII-FJXQXJEOSA-N | |
methyl (2S)-2-amino-3-(3,4-dihydroxyphenyl)propanoate;hydrochloride | |
10131132 | |
Powder or Crystalline Powder |
Safety and Handling
- L-3,4-Dihydroxyphenylalanine methyl ester hydrochloride, 97%
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.