missing translation for 'onlineSavingsMsg'
Learn More

L-3,4-Dihydroxyphenylalanine methyl ester hydrochloride, 95%, Thermo Scientific Chemicals

Catalog Number 11945491
Click to view available options
Quantity:
1 g
5 g
1421-65-4
C10H14ClNO4
247.68
WFGNJLMSYIJWII-UHFFFAOYNA-N
s-methyl 2-amino-3-3,4-dihydroxyphenyl propanoate hydrochloride, l-3,4-dihydroxyphenylalanine methyl ester hydrochloride, methyl l-dopa hydrochloride, melevodopa hydrochloride, unii-jfu1a8866v, l-dopa methyl ester hydrochloride, levodopa methyl ester hydrochloride, l-tyrosine, 3-hydroxy-, methyl ester, hydrochloride, dsstox_cid_25763, dsstox_rid_81105
10131132
[H+].[Cl-].COC(=O)C(N)CC1=CC=C(O)C(O)=C1
1421-65-4
C10H14ClNO4
247.68
WFGNJLMSYIJWII-UHFFFAOYNA-N
s-methyl 2-amino-3-3,4-dihydroxyphenyl propanoate hydrochloride, l-3,4-dihydroxyphenylalanine methyl ester hydrochloride, methyl l-dopa hydrochloride, melevodopa hydrochloride, unii-jfu1a8866v, l-dopa methyl ester hydrochloride, levodopa methyl ester hydrochloride, l-tyrosine, 3-hydroxy-, methyl ester, hydrochloride, dsstox_cid_25763, dsstox_rid_81105
10131132
[H+].[Cl-].COC(=O)C(N)CC1=CC=C(O)C(O)=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 94.2°C to 94.3°C
Color White
Elemental Analysis 45.58 to 51.40 % (C) Dry basis 5.32 to 5.99 % (N) Dry basis
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass Bottle
Quantity 1 g
Specific Rotation +9.0° to +11.0° (20°C, 589nm) (C=2 in methanol)
IUPAC Name methyl (2S)-2-amino-3-(3,4-dihydroxyphenyl)propanoate;hydrochloride
Formula Weight 247.68
Percent Purity 95%
Physical Form Crystalline Powder or Powder
Chemical Name or Material L-3,4-Dihydroxyphenylalanine methyl ester hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • L-3,4-Dihydroxyphenylalanine methyl ester hydrochloride, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'