missing translation for 'onlineSavingsMsg'
Learn More

L(+)-Arabinose, 99+%, Thermo Scientific Chemicals

Catalog Number 10023213
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
87-72-9
C5H10O5
150.13
SRBFZHDQGSBBOR-VVZXFQNISA-N
beta-d-arabinopyranose, 2r,3s,4r,5r-oxane-2,3,4,5-tetrol, pentopyranose #, .beta.-d-arabinopyranose, b-arabinopyranose, beta-arabinopyranose 9ci, methyl, a-d-arabinopyranoside, beta-d-arabinopyranose 9ci, 2r,3s,4r,5r-tetrahydro-2h-pyran-2,3,4,5-tetraol, 3s,2r,4r,5r-2h-3,4,5,6-tetrahydropyran-2,3,4,5-tetraol
25245970
(4S,5S)-oxane-2,3,4,5-tetrol
C1C(C(C(C(O1)O)O)O)O
87-72-9
C5H10O5
150.13
SRBFZHDQGSBBOR-VVZXFQNISA-N
beta-d-arabinopyranose, 2r,3s,4r,5r-oxane-2,3,4,5-tetrol, pentopyranose #, .beta.-d-arabinopyranose, b-arabinopyranose, beta-arabinopyranose 9ci, methyl, a-d-arabinopyranoside, beta-d-arabinopyranose 9ci, 2r,3s,4r,5r-tetrahydro-2h-pyran-2,3,4,5-tetraol, 3s,2r,4r,5r-2h-3,4,5,6-tetrahydropyran-2,3,4,5-tetraol
25245970
(4S,5S)-oxane-2,3,4,5-tetrol
C1C(C(C(C(O1)O)O)O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 155.0°C to 160.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99% min. (ELSD) (HPLC)
Packaging Plastic bottle
Quantity 100 g
Beilstein 31,32
Merck Index 10,783
Specific Rotation Condition +103.00 (20.00°C c=5,H2O,24H)
Specific Rotation + 103.00
Solubility Information (10% in water) Clear colorless
Formula Weight 150.13
Percent Purity 99+%
Physical Form Fine Crystalline Powder
Chemical Name or Material L(+)-Arabinose
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'