Learn More
L(+)-Arginine Hydrochloride, 98+%, ACROS Organics™
Brand: Acros Organics 105000025
Code : 29
Additional Details : CAS Number : 1119-34-2 Weight : 2.55000kg
Packaging | Plastic bottle |
---|---|
Quantity | 2.5kg |
Chemical Identifiers
1119-34-2 | |
210.67 | |
KWTQSFXGGICVPE-WCCKRBBISA-N | |
66250 | |
C(CC(C(=O)O)N)CN=C(N)N.Cl |
C6H14N4O2·HCl | |
MFCD00064550 | |
l-arginine hydrochloride, arginine hydrochloride, l-arginine hcl, l-arginine monohydrochloride, h-arg-oh.hcl, r-gene, arginine monohydrochloride, argamine, l-arginine, monohydrochloride, unii-f7lth1e20y | |
(2S)-2-amino-5-(diaminomethylideneamino)pentanoic acid;hydrochloride |
Specifications
L(+)-Arginine hydrochloride | |
0.5% max. (105°C, 2 hrs) | |
+22° (20°C c=8,6N HCl) | |
300ppm max | |
95% min. (c=10, H2O, 430nm) 1cm cell | |
226.0°C | |
1119-34-2 | |
C6H14N4O2·HCl | |
MFCD00064550 | |
l-arginine hydrochloride, arginine hydrochloride, l-arginine hcl, l-arginine monohydrochloride, h-arg-oh.hcl, r-gene, arginine monohydrochloride, argamine, l-arginine, monohydrochloride, unii-f7lth1e20y | |
KWTQSFXGGICVPE-WCCKRBBISA-N | |
(2S)-2-amino-5-(diaminomethylideneamino)pentanoic acid;hydrochloride | |
66250 | |
Crystals or Crystalline Powder |
Authentic | |
Plastic bottle | |
+ 22 | |
0.1% max. | |
White | |
2.5kg | |
98+% | |
H2NC(=NH)NH(CH2)3CH(NH2)CO2H·HCl | |
15,77 | |
Solubility in water: 75.1g/100g (20°C) | |
C(CC(C(=O)O)N)CN=C(N)N.Cl | |
210.67 | |
210.67 | |
98+% |