missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals L-Arginine monohydrochloride, 98+%
25.00 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
100 g
500 g
2500 g
Description
L-Arginine monohydrochloride is a precursor of nitric oxide synthesis that induces vasodilation, there by relaxing blood vessels. It is an important intermediate in the urea cycle and involved in the detoxification of nitrogenous wastes. It is commonly used in cell culture media and drug development.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
- L-Arginine is the left-handed enantiomer of the amino acid arginine, an essential amino acid in juvenile humans
- Arginine residues are common active site components in enzymes
- Arginine is involved in nitric acid production, and many biological processes depend on it
- Arginine plays a role in the immune system and in the detoxification of nitrogenous wastes
Application
- L-Arginine monohydrochloride is suitable for biochemistry and in vitro research
Specifications
Melting Point | 216°C (decomposition) |
Beilstein | 3631658 |
Merck Index | 14,780 |
Solubility Information | Soluble in water. Slightly soluble in hot alcohol. |
Formula Weight | 210.67 |
Percent Purity | ≥98% |
Density | 1.42 |
Quantity | 100 g |
Optical Rotation | +22° (c=5 in 5N HCl) |
Sensitivity | Hygroscopic |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'