missing translation for 'onlineSavingsMsg'
Learn More

L(-)-Borneol, 97%, Thermo Scientific Chemicals

Catalog Number p-72139
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
464-45-9
C10H18O
154.25
MFCD00003759,MFCD00066426,MFCD00066427
DTGKSKDOIYIVQL-NQMVMOMDSA-N
--borneol, borneol, l-borneol, 1s,2r,4s-1,7,7-trimethylbicyclo 2.2.1 heptan-2-ol, linderol, 1s-endo---borneol, borneo camphor, bornyl alcohol, sumatra camphor, dl-borneol
1201518
CHEBI:15394
(1S,3R,4S)-4,7,7-trimethylbicyclo[2.2.1]heptan-3-ol
CC1(C)[C@@H]2CC[C@]1(C)[C@H](O)C2
This item is not returnable. View return policy
464-45-9
C10H18O
154.25
MFCD00003759,MFCD00066426,MFCD00066427
DTGKSKDOIYIVQL-NQMVMOMDSA-N
--borneol, borneol, l-borneol, 1s,2r,4s-1,7,7-trimethylbicyclo 2.2.1 heptan-2-ol, linderol, 1s-endo---borneol, borneo camphor, bornyl alcohol, sumatra camphor, dl-borneol
1201518
CHEBI:15394
(1S,3R,4S)-4,7,7-trimethylbicyclo[2.2.1]heptan-3-ol
CC1(C)[C@@H]2CC[C@]1(C)[C@H](O)C2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 204°C to 208°C
Color White to Yellow
Flash Point 65°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Plastic bottle
Quantity 25 g
Beilstein 06,72
Merck Index 15,134
Specific Rotation Condition − 36.20 (20.00°C c=5,C2H5OH)
Specific Rotation -36.2
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
Formula Weight 154.25
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material L(-)-Borneol, 98%
Show More Show Less
compliance-icons
Product Identifier
  • L(-)-Borneol
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 2
Hazard Statement
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'