missing translation for 'onlineSavingsMsg'
Learn More

L-(-)-Malic acid, 99%, Thermo Scientific Chemicals

Catalog Number p-7042640
Click to view available options
Quantity:
100 g
500 g
2.5 kg
97-67-6
C4H6O5
134.087
MFCD00064213
BJEPYKJPYRNKOW-REOHCLBHSA-N
l-malic acid, l---malic acid, s-2-hydroxysuccinic acid, 2s-2-hydroxybutanedioic acid, l--malic acid, apple acid, --malic acid, l-apple acid, s-malic acid, s-2-hydroxybutanedioic acid
222656
CHEBI:30797
(2S)-2-hydroxybutanedioic acid
C(C(C(=O)O)O)C(=O)O
97-67-6
C4H6O5
134.087
MFCD00064213
BJEPYKJPYRNKOW-REOHCLBHSA-N
l-malic acid, l---malic acid, s-2-hydroxysuccinic acid, 2s-2-hydroxybutanedioic acid, l--malic acid, apple acid, --malic acid, l-apple acid, s-malic acid, s-2-hydroxybutanedioic acid
222656
CHEBI:30797
(2S)-2-hydroxybutanedioic acid
C(C(C(=O)O)O)C(=O)O

L-(-)-Malic acid is used as a food additive, Selective α-amino protecting reagent for amino acid derivatives. Versatile synthon for the preparation of chiral compounds including κ-opioid receptor agonists, 1α,25-dihydroxyvitamin D3 analogue, and phoslactomycin B.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-(-)-Malic acid is used as a food additive, Selective α-amino protecting reagent for amino acid derivatives. Versatile synthon for the preparation of chiral compounds including κ-opioid receptor agonists, 1α,25-dihydroxyvitamin D3 analogue, and phoslactomycin B.

Solubility
Soluble in water(363g/L).

Notes
Keep container tightly closed. Store away from oxidizing agents. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Melting Point 100°C to 106°C
Color White
Density 1.6
Flash Point 220°C (428°F)
Quantity 100 g
Beilstein 1723541
Merck Index 14,5707
Solubility Information Soluble in water(363g/L).
Optical Rotation −26° (c=5.5 in Pyridine)
Formula Weight 134.09
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material L-(-)-Malic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'