missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L(-)-Xylose, 99+%

Catalog Number p-4510255
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
609-06-3
C5H10O5
150.13
MFCD00151096
PYMYPHUHKUWMLA-UHFFFAOYNA-N
l-xylose, l +-xylose, l---xylose, 2s,3r,4s-2,3,4,5-tetrahydroxypentanal, aldehydo-l-xylose, l-xylo-pentose, l-xyl, xylose, l, linear l-xylose, l--xylose
95259
CHEBI:17979
OCC(O)C(O)C(O)C=O
This item is not returnable. View return policy
609-06-3
C5H10O5
150.13
MFCD00151096
PYMYPHUHKUWMLA-UHFFFAOYNA-N
l-xylose, l +-xylose, l---xylose, 2s,3r,4s-2,3,4,5-tetrahydroxypentanal, aldehydo-l-xylose, l-xylo-pentose, l-xyl, xylose, l, linear l-xylose, l--xylose
95259
CHEBI:17979
OCC(O)C(O)C(O)C=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 150.0°C to 152.0°C
Color White
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 99% min. (ELSD) (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 31,55
Specific Rotation Condition − 18.70 (24.00°C c=4,H2O,24h)
Specific Rotation − 18.70
Solubility Information Solubility in water: soluble. Other solubilities: soluble in pyridine and hot alcohol
IUPAC Name (2S,3R,4S)-2,3,4,5-tetrahydroxypentanal
Formula Weight 150.13
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material L(-)-Xylose
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'