missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Lab Vision™ Phosphate Buffered Saline, powder for a 1L 10X stock solution
Dilute this PBS tenfold with distilled water before use to make 1L of stock solution that acts as an ideal wash buffer for immunolabeling techniques.
Brand: Epredia™ AP-9009-10
Code : LD
Additional Details : Weight : 0.04536kg
Description
- For in vitro diagnostic use
- 1X working solution contains 0.14M Sodium chloride, 0.003M Potassium chloride, 0.002M Potassium phosphate, 0.01M Sodium phosphate
- Store at room temperature.
- Stable for up to three years
Specifications
1000 mL | |
10 X | |
Phosphate Buffered Saline |
Room temperature | |
Phosphate Buffered Saline (PBS) | |
Vitro Diagnostic |
For Research Use only