missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Laboratory Forceps

Manufactured from either PMP or POM for durability and resistance.

20.00 € - 33.55 €

Specifications

Product Type Forceps
View More Specs

Products 2
Product Code Brand Length (Metric) Tip Style Quantity (No. per Case) Price Quantity & Availability  
Product Code Brand Length (Metric) Tip Style Quantity (No. per Case) Price Quantity & Availability  
15281209
View Documents Promotion Details
BRAND™
113020
250 mm Rounded 5
33.55 €
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 04-06-2024
to see stock.
Add to basket
 
15251209
View Documents Promotion Details
BRAND™
112805
115 mm Pointed 10
20.00 €
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 07-05-2024
to see stock.
Add to basket
 
Description

Description

  • PMP forceps with pointed ends, autoclavable at 121°C, pack of 10
  • POM forceps with round ends, glass-fiber reinforced, pack of 5

Specifications

Specifications

Forceps
Videos
SDS
Documents

Documents

Product Certifications