missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Deutsch Neumann™ Laboratory Tubing
Provides flexibility, tear strength and resilience for general laboratory usage. Deutsch Neumann™ Laboratory Tubing is manufactured from red natural rubber.
Brand: Deutsch Neumann™ 3011217
Additional Details : Weight : 0.50000kg
Description
- Durometer 50 Shore A
- Good elastic memory
Specifications
ISO 9001:2008 | |
Natural Rubber | |
25 m | |
For Laboratory Use |
12 mm | |
17 mm | |
2.5 mm | |
Red |