missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lactose Monohydrate, Certified AR for Analysis, Fine Powder, Fisher Chemical™
Click to view available options
Quantity:
500 g
1 kg
Description
Lactose Monohydrate, Certified AR for Analysis, Fine Powder
Specifications
Melting Point | 219°C |
Color | White |
Packaging | Plastic powder jar |
Quantity | 500 g |
InChI Key | WSVLPVUVIUVCRA-ZEAJSSQNSA-N |
SMILES | C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O.O |
IUPAC Name | (3S,6R)-2-(hydroxymethyl)-6-[(3R,6R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol;hydrate |
Molecular Weight (g/mol) | 360.312 |
Formula Weight | 360.3g/mol |
Physical Form | Solid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'