Learn More
Thermo Scientific Chemicals Lactulose, 99+%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General description
• Lactulose is a disaccharide that is not absorbed during normal digestion
• May be capable of acting as a laxative and can capture NH3 (ammonia) in the colon
Applications
• Suitable for use in the research of bacterial metabolism and in vivo for physiology experiments
• Intended for biochemistry applications
Literature references
Guarino, MPL.; Altomare, A.; Emerenziani, S.; Di Rosa, C.; Ribolsi, M.; Balestrieri, P.; Iovino, P.; Rocchi, G.; Cicala, M. Mechanisms of Action of Prebiotics and Their Effects on Gastro-Intestinal Disorders in Adults. Nutrients. 2020, 12(4), 1037.
Specifications
Color | White |
pH | 3.0 to 7.0 (6% aq. soln.) |
Infrared Spectrum | Authentic |
Assay Percent Range | 99+% |
Packaging | Glass bottle |
Quantity | 5 g |
Specific Rotation Condition | − 47.00 (20.00°C c=5,H2O,12hr) |
Specific Rotation | -46.0° to -50.0° (20°C, 589nm) (c=5, H2O, ammonia) on anhydrous substance |
Formula Weight | 342.28 |
Percent Purity | ≥99.0% (on anhydrous substance) (HPLC) |
Show More |
RUO – Research Use Only