missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ Ladle Scoop

Manufactured from stainless steel. Bochem™ Ladle Scoop has a flat handle.

Brand:  Bochem™ 8742

Code : A0

Additional Details : Weight : 0.16200kg

 View more versions of this product

Product Code. 10028250

  • 26.60 € / Each
Estimated Shipment: 07-06-2024
to see stock.
Add to basket

Description

Description

  • Ladle scoop
  • Flat handle
  • Stainless steel
Specifications

Specifications

Ladle Scoop
Flat
Flat
315 mm
Stainless Steel (18/10)
220 mL
AISI 304
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers