missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lambda Light Chain Mouse, Clone: N10/2, enQuireBio™
Mouse Monoclonal Antibody
199.00 € - 485.00 €
Specifications
Antigen | Lambda Light Chain |
---|---|
Clone | N10/2 |
Concentration | 200μg/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15930258
|
EnquireBio
3535-MSM7-P1 |
100 μg |
485.00 €
100µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15920258
|
EnquireBio
3535-MSM7-P0 |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Lambda Light Chain | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
IGL | |
IgG1 κ | |
Protein A/G Purified | |
Cell Surface, cytoplasmic and secreted | |
293T, Raji or hPBL cells. Tonsil or Spleen |
N10/2 | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
3546 | |
Purified IgG from human serum | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
22.5kDa |