Learn More
Brady™ Laminated Polyester GHS Symbol Signs
Help ensure safety at work and maximize productivity with these GHS warning labels. Brady™ Laminated Polyester GHS Symbol Signs are available in a variety of sizes, with and without text. Standardized images reduce the risk of misunderstandings.
Brand: Brady™ 814039
Code : NEW
Additional Details : Weight : 0.20000kg
Description
- Self-adhesive diamond shape labels
- Pre-printed with GHS hazard symbols
- Laminated polyester, gloss finish black and red on white
Specifications
ISO 7010, RoHS compliant using EU Directive 2002/95/EC | |
Gloss | |
250 Pack | |
Permanent Acrylic | |
Pipe markers and safety signs | |
110°C | |
-40°C | |
GHS Symbol Sign | |
-40 °C-110 °C |
Red and Black on white | |
Polyester | |
Diamond | |
Material: Laminated Polyester | |
50mm | |
Corrosive | |
0.024mm | |
50mm |