Learn More
Lancer™ LancerClean LCD-P Laboratory Machine Detergent
A specially formulated, potassium hydroxide-based solution, LancerClean LCD-P can clean all types of items including glassware, surgical instruments and many other labware items.
Brand: Lancer™ 41010067
Code : 50
Additional Details : Weight : 1.00000kg Transport : UN number : 1719 Chem class : 8 Pack group : III
Description
- Made specifically for use in all high pressure washers
- Premixed for direct use from the bottle to the washer
- Dosing is automatic by peristaltic pump
- Manufactured with dedicated production lines and a double filtration system
- Capable of cleaning all different types of contaminants including laboratory soils, baked-on organic residues, petroleum and many more
- Phosphate-free and non-foaming
Specifications
Potassium hydroxide-based Cleaning Solution | |
All high pressure glassware washers | |
5 gal. |
Excellent cleaning results for all types of contaminants and standard laboratory soils | |
1 Ea. |
Safety and Handling
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Carcinogenicity Category 2
Hazard Statement
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H351-Suspected of causing cancer.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P264-Wash thoroughly after handling.
- P405-Store locked up.
Supplemental information
- MIXTURE LIST-Contains : Trisodium nitrilotriacetate, Sodium hydroxide