missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) bromide hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Catalog Number p-7049201
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
224183-16-8
Br3La
378.62
MFCD00798151
XKUYOJZZLGFZTC-UHFFFAOYSA-K
lanthanum iii bromide hydrate, acmc-1chtf, lanthanum bromide hydrate, lanthanum bromide-water 1/3/1
71317322
[Br-].[Br-].[Br-].[La+3]
This item is not returnable. View return policy
224183-16-8
Br3La
378.62
MFCD00798151
XKUYOJZZLGFZTC-UHFFFAOYSA-K
lanthanum iii bromide hydrate, acmc-1chtf, lanthanum bromide hydrate, lanthanum bromide-water 1/3/1
71317322
[Br-].[Br-].[Br-].[La+3]

Lanthanum(III) bromide is used as a scintillation material in certain applications. It is used in gamma spectroscopy radioisotope detection. It is used as a source of lanthanum in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Physical Form Crystalline
Quantity 10 g
Solubility Information Soluble in water.
IUPAC Name lanthanum(3+);tribromide;hydrate
Formula Weight 378.62 (Anhydrous)
Percent Purity 99.99%
Grade Reacton™
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Lanthanum(III) bromide hydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'